D-(+)-Fucose 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3615-37-0
:
Formula:C₆H₁₂O₅
MDL Number:MFCD00135603
Melting Pt:144…145 °C
Storage Temperature:Ambient
EINECS:222-792-9
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.5 % (K.F.)
- Solubility:(5 % in water) Clear colorless to slightly yellow
- HPLC:≥98.0 % (ELSD)
- Specific optical rotation:+74° to +78° (20°C, 589 nm) (c=2, water, NH3)
Specification Test Results
Frequently Bought Together