
Di(2-pyridyl) ketone 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:19437-26-4
:
Formula:C₁₁H₈N₂O
MDL Number:MFCD00006288
:184.20 g/mol
Melting Pt:53…55 °C
Flash Pt:>110 °C
EINECS:243-067-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Light yellow to beige to brown Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:49 to 54 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together