2,3,3-Trimethylindolenine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1640-39-7
:
Formula:C₁₁H₁₃N
MDL Number:MFCD00005724
:159.23 g/mol
Boiling Pt:228…229 °C (744 mmHg)
Density:0,99 g/cm³
Flash Pt:93 °C
EINECS:216-685-6
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear yellow to red-brown Liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together