Forskolin, Millipore®
Supplier: Merck Millipore (Calbiochem)
Certificates
About this item
Synonyms: (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-6,10,10b-trihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-3-vinyldodecahydro-1H-benzo[f]chromen-5-yl acetate
CAS Number:66575-29-9
:
Formula:C₂₂H₃₄O₇
MDL Number:MFCD00082317
Molecular Weight:410.51 g/mol
EINECS:266-410-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together