Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Carbamazepine 1000 µg/ml in methanol, Cerilliant
Organic reference standard, Carbamazepine 1000 µg/ml in methanol, Cerilliant
Catalog # PROOCERC-053
CAS Number:  
Organic reference standard, Carbamazepine 1000 µg/ml in methanol, Cerilliant
Catalog # PROOCERC-053
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    298-46-4
  • Description:
    Carbamazepine 1,000 µg/ml in methanol
  • Formula:
    C₁₅H₁₂N₂O
  • Description:
    Carbamazepine

Specifications

About this item