Organic reference standard, Hydrochlorothiazide 1,000 µg/ml in methanol, Cerilliant
Catalog # PROOCERH-001
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:58-93-5
- Description:Hydrochlorothiazide 1,000 µg/ml in methanol
- Formula:C₇H₈ClN₃O₄S₂
- Description:Hydrochlorothiazide