Order Entry
Ireland
ContactUsLinkComponent
Tubing connectors, straight, HDPE
Tubing connectors, straight, HDPE
Catalog # 229-7486
Supplier:  SEMADENI
undefined
Tubing connectors, straight, HDPE
Catalog # 229-7486
Supplier:  SEMADENI
Specifications
  • Hole Ø:
    20 mm
  • Tubing int.Ø:
    25 - 27 mm
  • Pk:
    1
Specifications
About this item
Tubing connector, made of HDPE, with taper ends.
  • Good temperature resistance
  • Good chemical resistance