Paper labels, Freeza-Seal
Catalog # 562-562
Supplier: JENCONS SCIENTIFIC
Specifications
- Printer model:
- Printing method:
- Colour:White/blue
- Description:Biohazard labels
- Dimensions:19 mm
- Material:Paper
- Environmentally Preferable:
- Packed:1000 labels/pack
- Pk:1
Specifications
About this item
Peel-off sheets of self-adhesive Freeza-Seal miniature labels.
- Adheres to all containers to approx. −40 °C
- Round, 19 mm diameter