Order Entry
Ireland
ContactUsLinkComponent
Leak diverter for roofs, PIG®
  NEWPTLS564-YW
 :  NEW PIG
undefined
Leak diverter for roofs, PIG®
  NEWPTLS564-YW
 :  NEW PIG
  • Description:
    Leak diverter with spring loaded aluminium pole, vinyl coated tarpulin, yellow
  • Height:
    279,4 mm
  • Pk:
    1
Leak diverter with telescopic, aluminium pole which extends into place to install the leak diverter up to 6 metres in the air without a ladder.
  • Tension mounted pole with rubber cups secures pole to floor and ceiling
  • Makes it easy for one person to install or move the diverter
  • Non permeable tarpaulin is vinyl coated for strength and durability
  • Resists punctures, UV and mildew for long life
 : Hose sold separately.