Order Entry
Ireland
ContactUsLinkComponent
Spill station sign, PIG®
Spill station sign, PIG®
Catalog # NEWPSGN303
Supplier:  NEW PIG
undefined
Spill station sign, PIG®
Catalog # NEWPSGN303
Supplier:  NEW PIG

Specifications

  • Description:
    Spill station, yellow/black, 0,39 kg
  • Dimensions:
    380×200 mm
  • Material:
    PETG
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

These 3D wall signs make emergency spill kits easy to find from any angle. Faster to find means faster to respond.

  • Protrudes from wall for higher visibility
  • Even from a distance, helps indicate the location of spill kits and supplies that may be obscured in their immediate area
  • Highly-visible with high-contrast message on both sides

The signs keep your facility looking professional while providing clear safety information.