Order Entry
Ireland
ContactUsLinkComponent
Cy™ fluorescent amidites
Cy™ fluorescent amidites
Supplier:  Cytiva
Certificates
undefined
Cy™ fluorescent amidites
Supplier:  Cytiva

Specifications

  • Physical form:
    Solid
  • Storage conditions:
    Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, away from incompatible materials. Keep container tightly closed and sealed until ready for use
  • Solubility:
    Soluble in cold water and hot water
  • Stability:
    Stable

Specifications

Related Information

Frequently Bought Together