Order Entry
Ireland
ContactUsLinkComponent
Columns, HiPrep™ Capto Q
Columns, HiPrep™ Capto Q
Catalog # 28-9375-48
Supplier:  Cytiva
Columns, HiPrep™ Capto Q
Catalog # 28-9375-48
Supplier:  Cytiva
Specifications
  • Description:
    HiPrep™ Capto Q column
  • Bed dimensions:
    16×100 mm
  • Pk:
    1