Specialty solid phase extraction cartridges, Bond Elut™ Carbon/NH₂
: VARIAN
These specialty solid phase extraction cartridges provide higher retention of small organics, including those that are too polar to retain on C18 or polymeric SPE.
- Typical matrices include organic plant and tissue extracts
- Removal of chlorophyll and other pigments leads to fewer chromatographic or mass interferences
- Broader retention and easier elution of analytes across the polarity range, for improved multi-residue analysis
These cartridges are packed with ultra-pure graphitised carbon particles that have been optimised for the absorption of pigments in food, fruits and vegetables, and small organic residues in waste water.
Frequently Bought Together