Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Supplier: Thermo Fisher Scientific
Certificates
About this item
Aminopeptidase B inhibitor. Also inhibits leucine aminopeptidase.
CAS No.: 58970-76-6 Molecular Formula: C16H24N2O4 Formula Weight: 308.37 Physical Form: Powder Appearance: White Melting Point: 233-236° MDL No.: MFCD00083262
Aminopeptidase B inhibitor. Also inhibits leucine aminopeptidase
Specifications
- CAS No.:58970-76-6
- Storage conditions:Ambient
- Form:Powder
- MDL No.:MFCD00083262
- Protein synonyms:Ubenimex|Bestatin-S-R-S-N-3-amino-2-hydroxy-4-phenylbutyroyl-L-leucine|N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine
- Protein/peptide name:Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
- Molecular weight:308,4 kDa
- Molecular formula:C₁₆H₂₄N₂O₄
Specifications
Related Information
Frequently Bought Together