Order Entry
Ireland
ContactUsLinkComponent
Benzyl alcohol ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Benzyl alcohol ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Catalog # 80708-1L
CAS Number:  
undefined
Benzyl alcohol ≥99.9% (by GC), CHROMASOLV™ for headspace gas chromatography, Riedel-de Haën™
Catalog # 80708-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Refractive index (n 20/D):
    1.538 - 1.541
  • Water (Karl Fischer):
    Max. 0.02 %
  • Acetophenone (GC):
    Max. 0.01 %
  • Benzaldehyde (GC):
    Max. 0.01 %
  • Suitability for the GC Headspace:
    complying
  • Vapor Pressure:
    3.75 mmHg ( 77 °C)
  • Autoignition Temperature:
    817 °F
  • Vapor Density:
    3.7 (vs air)

Specification Test Results

About this item

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol
 :
 :108.14 g/mol
Merck Index:13,01126
Boiling Pt:205,3 °C (1.013 hPa)
Melting Pt:-15,4 °C
Melting Pt:–15,4 °C
Density:1,045 g/cm³ (20 °C)
Flash Pt:100,4 °C
Storage Temperature:Ambient
EINECS:202-859-9
 :GHS07
CAS Number:100-51-6
Formula:C₆H₅CH₂OH
MDL Number:MFCD00004599