Order Entry
Ireland
ContactUsLinkComponent
4-(Trifluoromethylthio)benzyl bromide ≥97%
Corrosive
4-(Trifluoromethylthio)benzyl bromide ≥97%
Catalog # L19205.03
CAS Number:  
Corrosive
4-(Trifluoromethylthio)benzyl bromide ≥97%
Catalog # L19205.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:21101-63-3
 :
Formula:C₈H₆BrF₃S
MDL Number:MFCD00129189
 :271.10 g/mol
Boiling Pt:115…118 °C (13 mmHg)
Melting Pt:53…56 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05