Chemical Resistant Overalls, 4535
Supplier: 3M
Certificates
About this item
These disposable overalls made of PP with a PE laminate provide an excellent barrier against dry particles and certain limited liquid chemical splashes.
- Elasticated waist and ankles for freedom of movement and knitted cuffs for extra comfort
- Extra large breathable back panel (SMMMS) for reduced heat stress and increased comfort
- Two-way zip, hood and sealable storm flap for enhanced protection
- Low linting properties and anti-static coating
Typical light duty applications may include paint spraying, industrial clean-up and maintenance, industrial and chemical processing, building cleaning, insulation laying, pesticide handling/agriculture (particulate aerosols only), powder coating and pharmaceutical manufacturing.
Colour: White/blue
Certifications: Type 5/6, EN 1149-5, EN 1073-2:2002; Cat. III.
Specifications
- Catalog No:
- 113-6296
- 113-6299
- 113-6295
- 113-6297
- Model:
- 4535
- 4535
- 4535
- 4535
- Anti-static:
- +
- +
- +
- +
- Garment application:
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial
- Chemical protection, Laboratory, Industrial