

Trifluoroacetic acid for biotechnology
0993-500ML
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
:
:114.02 g/mol
:13,09754
:71,8 °C (1013 hPa)
:–15 °C
:1,490 g/cm³ (20 °C)
:Ambient
:200-929-3
:2699
:8,I
:GHS05
:GHS07
:76-05-1
:C₂HF₃O₂
:MFCD00004169