3'-(Trifluoromethyl)acetanilide
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:351-36-0
:
Formula:C₉H₈F₃NO
MDL Number:MFCD00000383
:203.16 g/mol
Boiling Pt:287 °C (1013 hPa)
Melting Pt:107…108 °C
EINECS:206-512-2
Frequently Bought Together