

Acetaldehyde oxime (mixture of syn and anti) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:107-29-9
:
Formula:C₂H₅NO
MDL Number:MFCD00002124
:59.07 g/mol
Melting Pt:44…46 °C
Density:0,96 g/cm³
Flash Pt:38 °C
Storage Temperature:Refrigerator
EINECS:203-479-6
UN:2332
ADR:3,III
:GHS06
:GHS02
Specification Test Results
- Appearance:Clear colorless to white Crystals or low melting solid or liquid
- Infrared spectrum:Conforms
- Refractive index:1.4230 to 1.4270 (20°C, 589 nm)
- GC:≥98.5 %
- Color scale:≤10 APHA
Specification Test Results
Frequently Bought Together