2,4,6-Triisopropylbenzenesulphonamide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:105536-22-9
:
Formula:C₁₅H₂₅NO₂S
MDL Number:MFCD00051975
:283.44 g/mol
Melting Pt:113…116 °C
Storage Temperature:Ambient
Related Information
Frequently Bought Together