
Methyl-4-aminosalicylate ≥97%
Catalog # 45520.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
CAS Number:4136-97-4
:
Formula:C₈H₉NO₃
MDL Number:MFCD00088091
:167.18 g/mol
EINECS:223-958-3
:GHS07
Storage Temperature:Ambient