

2-Acrylamido-2-methylpropanesulphonic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15214-89-8
:
Formula:C₇H₁₃NO₄S
MDL Number:MFCD00007522
:207.25 g/mol
Melting Pt:195 °C (dec.)
Flash Pt:160 °C
EINECS:239-268-0
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder or chips and/or chunks
- Infrared spectrum:Conforms
- Titration with NaOH:≥96.0 %
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together