4'-Cyano-2'-(trifluoromethoxy)acetanilide
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:175278-19-0
:
Formula:C₁₀H₇F₃N₂O₂
MDL Number:MFCD00204174
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together