Organic reference standard, Tolbutamide 1,000 µg/ml in acetonitrile, Cerilliant
PROOCERT-036
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:64-77-7
- Description:Tolbutamide 1,000 µg/ml in acetonitrile
- Formula:C₁₂H₁₈N₂O₃S
- Description:Tolbutamide