Organic reference standard, Estrone 10 µg/ml in methanol
Catalog # PROONE9253
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:53-16-7
- Description:Estrone 10 µg/ml in methanol
- Formula:C₁₈H₂₂O₂
- Description:Estrone