Hydrochloric acid 4 mol/l, EMPROVE® EXPERT for biopharmaceutical production
Supplier: MERCK PRODUCTION CHEMICALS
Certificates
About this item
Synonyms: Hydrochloric acid solution, Muriatic acid
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
Density:1.06 g/cm³ (20 °C)
Molecular Weight:36.46 g/mol
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
ADR:8
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together