Order Entry
Ireland
ContactUsLinkComponent
Disinfectants, High Level Laboratory Surface Disinfectant, Distel
Disinfectants, High Level Laboratory Surface Disinfectant, Distel
Supplier:  TRISTEL SOLUTIONS
Certificates
undefined
Disinfectants, High Level Laboratory Surface Disinfectant, Distel
Supplier:  TRISTEL SOLUTIONS
Certificates
Specifications
  • Catalog No:
  • 141-0935
  • Model:
  • Distel for labs
  • Bactericidal activity:
  • Pseudomonas aeruginosa, Campylobacter spp, Staphylococcus spp, Streptococcus spp, Leptospira spp, Escherichia coli, Enterococcus spp , Klebsiella spp, Salmonella spp, Listeria monocytogenes, Legionella spp, Proteus vulgaris, Rhodococcus spp, Serratia spp Mycobacterium tuberculosis, Mycobacterium terrae, Mycobacterium avium
  • Fungicidal activity:
  • Aspergillus niger, Trichophyton spp, Candida spp
  • Virucidal activity:
  • Coronavirus (SARS), Adenovirus type 5, Influenza
  • pH:
  • 4.5 - 6.5