About this item
Thick natural latex rubber gloves, providing greater protection for heavy-duty applications.
Advanced protection: AlphaTec® 87-208 heavy-duty chemical-resistant gloves are 50% thicker (0,75 mm/29,5 mil) than standard flock-lined alternatives, withstanding various fats, ketones, salts, detergents, alcohols, and alkalis
Enhanced cleanliness and durability: They undergo a chlorination treatment that cleanses and hardens their external surface
Elevated comfort: Designed with 100% cotton flocking, these heavy-duty protective gloves absorb moisture and sweat, for greater wearer comfort
Assured quality: In addition, these AlphaTec® gloves achieve an AQL of 1.5
Recommended for: Maintenance of plant and machines, chemical handling, janitorial use
Primary industries: Construction, infrastructure maintenance, janitorial/sanitation, metal fabrication, chemical, utilities, warehousing
Specifications
- Catalog No:
- ANSE87-208/7
- ANSE87-208/8
- ANSE87-208/9
- ANSE87-208/10
- ANSE87-208/11
- Latex/Latexmentes:
- Latex
- Latex
- Latex
- Latex
- Latex
- Felületi kiképzés:
- Honeycomb
- Honeycomb
- Honeycomb
- Honeycomb
- Honeycomb
- Kézelő típusa:
- Egyenes
- Egyenes
- Egyenes
- Egyenes
- Egyenes
- Tenyérrész vastagsága:
- 0,75 mm
- 0,75 mm
- 0,75 mm
- 0,75 mm
- 0,75 mm
- EN előírás:
- EN 388:2016 X121X, EN ISO 21420:2020, EN ISO 374-1:2016 Type A AKLOPS, EN ISO 374-5 VIRUS
- EN 388:2016 X121X, EN ISO 21420:2020, EN ISO 374-1:2016 Type A AKLOPS, EN ISO 374-5 VIRUS
- EN 388:2016 X121X, EN ISO 21420:2020, EN ISO 374-1:2016 Type A AKLOPS, EN ISO 374-5 VIRUS
- EN 388:2016 X121X, EN ISO 21420:2020, EN ISO 374-1:2016 Type A AKLOPS, EN ISO 374-5 VIRUS
- EN 388:2016 X121X, EN ISO 21420:2020, EN ISO 374-1:2016 Type A AKLOPS, EN ISO 374-5 VIRUS