Order Entry
Hungary
Orders LinkContactUsLinkComponent
Monkey Eotaxin ELISA Kit

Monkey Eotaxin ELISA Kit

Monkey Eotaxin ELISA Kit, 384 tests
Catalog # ABCAAB317549-1X384
New Product
Supplier:   Abcam
Monkey Eotaxin ELISA Kit
Catalog # ABCAAB317549-1X384
Supplier:  Abcam
New Product
CAS Number:  
Monkey Eotaxin ELISA Kit
Monkey Eotaxin ELISA Kit

Monkey Eotaxin ELISA Kit, 384 tests

CAS Number:  
Price
814 360 Ft
Log in for availability
1 * 1 Tests
Product Details & Documents

Monkey Eotaxin ELISA kit is a single-wash 90-min Simplestep used to quantify Monkey Eotaxin with a sensitivity of 12.913 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 384-well plate for higher throughput.

Specifications

Specifications

  • Description:
    Monkey Eotaxin ELISA Kit
  • Tests per kit:
    384 tests
  • Primary antibody reactivity:
    Monkey
  • Sample type:
    Serum, EDTA Plasma, Citrate plasma
  • Detection range:
    39,063 - 2500 pg/ml
  • Storage temperature:
    4 °C
  • Assay Type:
    Sandwich
  • Time to Results:
    1 hr 30 min
  • Cat. No.:
    ABCAAB317549-1X384
  • Assay duration:
    One step
  • Detection method:
    Colorimetric
  • Sensitivity:
    12,913 pg/ml
  • Format:
    Pre-coated
  • Target protein:
    Eotaxin
  • Regulatory status:
    RUO

Specifications

Ajánlott termékek

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.