Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
3 COMPONENTS: 100MG/L EACH 1 * 5 Ampoul
Catalog # CMPLRD0299251.5
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Expand image
3 COMPONENTS: 100MG/L EACH 1 * 5 Ampoul
Catalog # CMPLRD0299251.5
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Potassium hydroxide ≥85%, pellets, GPR RECTAPUR®
Supplier: VWR Chemicals
VISOCOLOR® Colorimetric Test, Chromium
Supplier: MACHEREY-NAGEL
Sodium hydroxide 32% in aqueous solution, AnalaR NORMAPUR® analytical reagent, for nitrogen analysis
Supplier: VWR Chemicals
Test strips, nitrite, Dosatest
Supplier: VWR Chemicals
VWR® Rota®, pH Indicator Paper, 1 to 14
Supplier: VWR Chemicals
pH indicator test strips, pH-Fix
Supplier: MACHEREY-NAGEL
Sodium hypochlorite (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Test strips, water hardness, Dosatest
Supplier: VWR Chemicals
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
Supplier: Hach