Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Rendelés felvitel
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]1 COMPONENT: P (PO43-) 2MG/L IN H2O 1 * 100 mL
Catalog # CMPLRM065176L1
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Expand image
[EN]1 COMPONENT: P (PO43-) 2MG/L IN H2O 1 * 100 mL
Catalog # CMPLRM065176L1
Supplier:
CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Nátrium hidroxid 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Hangyasav ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Nátrium hidroxid 32% in aqueous solution, AnalaR NORMAPUR® analytical reagent, for nitrogen analysis
Supplier: VWR Chemicals
Ammóniaoldat 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Test kits, photometric, phosphate, LCK cuvette tests
Supplier: Hach