This Triglyceride Assay Kit is a quantitative assay kit for measuring Triglyceride in serum, plasma, animal and plant tissues, cells , cell supernatant, and bacteria. 1 * 96 Tests
Catalog # ANTIA319752-96
Supplier: Antibodies.com