Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
This Tannin Assay Kit is a quantitative assay kit for measuring Tannin in plant tissues and liquid samples such as juice and honey. 1 * 96 Tests
Catalog # ANTIA319743-96
Supplier:
Antibodies.com
New Product
Expand image
This Tannin Assay Kit is a quantitative assay kit for measuring Tannin in plant tissues and liquid samples such as juice and honey. 1 * 96 Tests
Catalog # ANTIA319743-96
Supplier:
Antibodies.com
New Product
Frequently Bought Together
VWR®, Petri Dish, PS
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cell culture flasks, Falcon®
Supplier: Corning
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Transfer pipettes
Supplier: RATIOLAB
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection