Skip to Header
Skip to Main Content
Skip to Footer
Bővített
Bővített
SDS
Tanúsítványok
Gyártói adatok
0
0 Ft
Order Entry
Login
/
Regisztráció
Products
Services
Digital Solutions
Applications
Support
About
Gyors rendelés
Megrendelések állapota
Saját fiók
Kosár
Hungary
Gyors rendelés
Megrendelések állapota
Saját fiók
Kosár
ContactUsLinkComponent
This Tannase Activity Assay Kit is a quantitative assay kit for measuring Tannase activity in plant tissues, fungi, and bacteria. 1 * 48 Tests
Catalog # ANTIA319742-48
Supplier:
Antibodies.com
New Product
Expand image
This Tannase Activity Assay Kit is a quantitative assay kit for measuring Tannase activity in plant tissues, fungi, and bacteria. 1 * 48 Tests
Catalog # ANTIA319742-48
Supplier:
Antibodies.com
New Product
Frequently Bought Together
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Cell culture flasks, Nunc™
Supplier: Thermo Fisher Scientific
VWR®, Sampling Spoons
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Transfer pipettes
Supplier: RATIOLAB
VWR® Ultra-High Performance Centrifuge Tubes
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection