0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
This Soil Phosphate Assay Kit is a quantitative assay kit for measuring Phosphate in soil. 1 * 96 Tests
ANTIA319735-96
:
Antibodies.com
New Product
This Soil Phosphate Assay Kit is a quantitative assay kit for measuring Phosphate in soil. 1 * 96 Tests
ANTIA319735-96
:
Antibodies.com
New Product
Frequently Bought Together
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
VWR® Premium Line, Serological Pipettes
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Serological pipettes
: Greiner Bio-One
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Cell Strainers
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF