Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
This Serum Sodium Assay Kit is a quantitative assay kit for measuring Sodium in serum. 1 * 96 Tests
Catalog # ANTIA319732-96
Supplier:
Antibodies.com
New Product
Expand image
This Serum Sodium Assay Kit is a quantitative assay kit for measuring Sodium in serum. 1 * 96 Tests
Catalog # ANTIA319732-96
Supplier:
Antibodies.com
New Product
Frequently Bought Together
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
Centrifuge tubes, 15 ml, Corning®
Supplier: Corning
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR® Blender Bags
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
Centrifuge tubes, Falcon®
Supplier: Corning