Order Entry
Hungary
ContactUsLinkComponent
This Mitochondrial Complex IV Activity Assay Kit is a quantitative assay kit for measuring Mitochondrial Complex IV activity in anim al tissue , plant tissue and cell samples. 1 * 96 Tests
Catalog # ANTIA319702-96
Supplier:  Antibodies.com
New Product
This Mitochondrial Complex IV Activity Assay Kit is a quantitative assay kit for measuring Mitochondrial Complex IV activity in anim al tissue , plant tissue and cell samples. 1 * 96 Tests
Catalog # ANTIA319702-96
Supplier:  Antibodies.com