LIVE AND DEAD CELL DOUBLE STAINING KIT 1 * 2.000 Tests
Catalog # ANTIA319650-2000
Supplier: Antibodies.com
New Product
LIVE AND DEAD CELL DOUBLE STAINING KIT 1 * 2.000 Tests
Catalog # ANTIA319650-2000
Supplier: Antibodies.com
New Product
Frequently Bought Together
Filter plates, 96-well, for multiplexing, AcroPrep™ Advance
Supplier: Cytiva (Formerly Pall Lab)
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Nalgene®, Biotainer® Bottles
Supplier: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
Nalgene®, Biotainer® Bottles
Supplier: Thermo Fisher Scientific
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Heat sealing foil, PierceASeal Foil™
Supplier: iST Scientific Ltd
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION
Bottle hdpe 250ml 18mg sod thio 0.125ml 1 * 25 items
Supplier: LAB3 OWN PRODUCTION
VWR®, Roller Bottles
Supplier: VWR Collection
Nunc™ Internally-Threaded Universal Tubes and Rack System
Supplier: Thermo Fisher Scientific
PCR plates, 384-well, Thermo-Fast® 384
Supplier: Thermo Fisher Scientific
Blotting transfer papers
Supplier: Ahlstrom-Munksjö
Spare Lid of Ice Pan 4L, colour option, RED, BLUE, GREEN, BLACK 1 * 1 items
Supplier: VWR Collection