Order Entry
Hungary
ContactUsLinkComponent
This Caspase-6 Assay Kit (Colorimetric) is a quantitative assay kit for measuring Caspase-6 activity in animal tissues and cells. 1 * 20 Tests
  ANTIA319628-20
New Product
This Caspase-6 Assay Kit (Colorimetric) is a quantitative assay kit for measuring Caspase-6 activity in animal tissues and cells. 1 * 20 Tests
  ANTIA319628-20