0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
This Caspase-4 Assay Kit (Colorimetric) is a quantitative assay kit for measuring Caspase-4 activity in animal tissues and cells. 1 * 100 Tests
ANTIA319627-100
:
Antibodies.com
New Product
This Caspase-4 Assay Kit (Colorimetric) is a quantitative assay kit for measuring Caspase-4 activity in animal tissues and cells. 1 * 100 Tests
ANTIA319627-100
:
Antibodies.com
New Product
Frequently Bought Together
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
Laboratory trays
: KARTELL
VWR®, Cryoboxes, Cardboard, 136x136 mm
: VWR Collection
VWR®, Sample Dippers
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
Microcentrifuge tubes, 0,6/1,5/1,7/2,0/5,0 ml, SnapLock, Axygen®
: Corning
VWR®, Ice Pans, with or without Lid
: VWR Collection
VWR®, Conical Sample Containers, with Snap-On Lids
: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Centrifuge tubes, sterile, polypropylene, Corning®
: Corning
VWR®, Real-Time PCR Plates, 96-Well and 384-Well
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection