0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
This Caspase-3 Assay Kit (Colorimetric) is a quantitative assay kit for measuring Caspase-3 activity in animal tissues and cells. 1 * 100 Tests
ANTIA319626-100
:
Antibodies.com
New Product
This Caspase-3 Assay Kit (Colorimetric) is a quantitative assay kit for measuring Caspase-3 activity in animal tissues and cells. 1 * 100 Tests
ANTIA319626-100
:
Antibodies.com
New Product
Frequently Bought Together
VWR®, Conical Sample Containers, with Snap-On Lids
: VWR Collection
Serological pipettes, Stripette™
: Corning
VWR®, Microcentrifuge Tubes, High Performance
: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
: EPPENDORF
VWR®, Petri Dish, PS
: VWR Collection
VWR®, Transfer Pipettes, Graduated
: VWR Collection
Centrifuge tubes
: Brand
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Reagent/centrifuge tube with screw cap
: Greiner Bio-One
Media bottles, square, sterile, Nalgene®
: Thermo Fisher Scientific
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Sample Dippers
: VWR Collection
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection