Skip to Header
Skip to Main Content
Skip to Footer
Bővített
Bővített
SDS
Tanúsítványok
Gyártói adatok
0
0 Ft
Order Entry
Login
/
Regisztráció
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
This Caspase-2 Assay Kit (Colorimetric) is a quantitative assay kit for measuring Caspase-2 activity in animal tissues and cells. 1 * 20 Tests
Catalog # ANTIA319625-20
Supplier:
Antibodies.com
New Product
Expand image
This Caspase-2 Assay Kit (Colorimetric) is a quantitative assay kit for measuring Caspase-2 activity in animal tissues and cells. 1 * 20 Tests
Catalog # ANTIA319625-20
Supplier:
Antibodies.com
New Product
Frequently Bought Together
Serological pipettes, Stripette™
Supplier: Corning
Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
VWR®, Strips of PCR Tubes and Caps
Supplier: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Supplier: Corning
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Sampling Spoons
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
Supplier: VWR Collection