Order Entry
Hungary
ContactUsLinkComponent
This Caspase-1 Assay Kit (Colorimetric) is a quantitative assay kit for measuring Caspase-1 activity in animal tissues and cells. 1 * 50 Tests
Catalog # ANTIA319624-50
Supplier:  Antibodies.com
New Product
This Caspase-1 Assay Kit (Colorimetric) is a quantitative assay kit for measuring Caspase-1 activity in animal tissues and cells. 1 * 50 Tests
Catalog # ANTIA319624-50
Supplier:  Antibodies.com