Rabbit polyclonal antibody to LC3B. 1 * 100 µl
ANTIA88451-100
New Product
Frequently Bought Together
VWR® Premium Line, Serological Pipettes
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR® Ultra-High Performance Centrifuge Tubes
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Cell Strainers
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Serological pipettes, Stripette™
: Corning
VWR®, Conical Sample Containers, with Snap-On Lids
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection