Rabbit polyclonal antibody to Ku80. 1 * 100 µl
ANTIA91434-100
New Product
Frequently Bought Together
VWR®, Microcentrifuge Tubes and Screw Caps
: VWR Collection
VWR®, Spatulas
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
Seals for ALPS™ 50 V heat sealer
: THERMO MATRIX TECHNOLOGIES
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
: EPPENDORF
Cell culture flasks, Nunc™
: Thermo Fisher Scientific
Nalgene®, Biotainer® Bottles
: Thermo Fisher Scientific
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
VWR®, Ultra-Clear Films for qPCR
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Blotting Pads, Grade 707
: VWR Collection
Microplates, 96-well
: Greiner Bio-One