Rabbit polyclonal antibody to PANDER. 1 * 100 µl
ANTIA89081-100
New Product
Frequently Bought Together
Sampling spatula, Sterileware®, SP Bel-Art
: Bel-Art Products, a Part of SP
VWR®, Strips of PCR Tubes and Caps
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
: EPPENDORF
Seals for ALPS™ 50 V heat sealer
: THERMO MATRIX TECHNOLOGIES
VWR®, Cryoboxes, Cardboard, 133×133 mm
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Cryo Boxes
: VWR Collection
Inoculating needles and loops, Nunc™
: Thermo Fisher Scientific