Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 Ft
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Hungary
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Rabbit polyclonal antibody to FADD. 1 * 100 µl
ANTIA89025-100
:
Antibodies.com
New Product
Rabbit polyclonal antibody to FADD. 1 * 100 µl
ANTIA89025-100
:
Antibodies.com
New Product
Frequently Bought Together
VWR® Serological Pipette, Standard Line
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Petri Dish, PS
: VWR Collection
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR® Water Sampling Bottles, with Tamper Evident Cap
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection