0 Ft
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Hungary
ContactUsLinkComponent
Rabbit polyclonal antibody to Smad2. 1 * 100 µl
ANTIA80517-100
:
Antibodies.com
New Product
Rabbit polyclonal antibody to Smad2. 1 * 100 µl
ANTIA80517-100
:
Antibodies.com
New Product
Frequently Bought Together
VWR® Blender Bags
: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
: Corning
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
VWR® PCR Tubes, 0,2 ml
: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR® Standard Line Transfer Pipettes
: VWR Collection
VWR®, Sampling Spoons
: VWR Collection