LAMBDA PROTEIN PHOSPHATASE (FUNCTIONAL) 1 * 20 KU
Catalog # ANTIA461-20
Supplier: Antibodies.com
New Product
LAMBDA PROTEIN PHOSPHATASE (FUNCTIONAL) 1 * 20 KU
Catalog # ANTIA461-20
Supplier: Antibodies.com
New Product
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
VWR®, Centrifuge Tubes, Conical Bottom, PP
Supplier: VWR Collection
VWR®, Cryoboxes, Cardboard, 136x136 mm
Supplier: VWR Collection
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
Measuring scoops
Supplier: Brand
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW
Multi-layer cell culture flasks, Falcon® Multi-Flask
Supplier: Corning
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Supplier: STERILIN
Blotting transfer papers
Supplier: Ahlstrom-Munksjö
Detectable spoons, SteriPlast®
Supplier: BURKLE
BOTTLE PET 250ML SQUARE T/E GREEN CAP 1 * 200 items
Supplier: LAB3 OWN PRODUCTION